• USA Home
  • 8.14963 - 4-Bromo-2-fluorobiphenyl

EMAIL THIS PAGE TO A FRIEND
8.14963 EMD Millipore

4-Bromo-2-fluorobiphenyl

4-Bromo-2-fluorobiphenyl for synthesis. CAS 41604-19-7, molar mass 251.1 g/mol., for synthesis

  • CAS Number 41604-19-7

  • Linear Formula C₁₂H₈BrF

  • Molecular Weight 251.1 g/mol

  •  EC Number 255-453-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Glass bottle (8149630050)
vapor pressure   0.04 hPa (20 °C)
density   1.4 g/cm3 (20 °C)
flash point   113 °C
mp   38.5 °C
LD 50 oral   LD50 Rat > 1000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 38 °C
  Melting range (upper value): ≤ 41 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 3152 , 9, II
Declaration (transport by air) IATA-DGR 
UN 3152 , 9, II
Declaration (transport by sea) IMDG-Code 
UN 3152 , 9, II, Marine Pollutant: P
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?