• USA Home
  • 8.14966 - 1-Bromo-3,4,5-trifluorobenzene

EMAIL THIS PAGE TO A FRIEND
8.14966 EMD Millipore

1-Bromo-3,4,5-trifluorobenzene

1-Bromo-3,4,5-trifluorobenzene for synthesis. CAS 138526-69-9, molar mass 210.98 g/mol., for synthesis

  • CAS Number 138526-69-9

  • Linear Formula C₆H₂BrF₃

  • Molecular Weight 210.98 g/mol

  •  EC Number 418-480-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8149660010)
bp   140 °C (1006 hPa)
vapor pressure   8.65 hPa (25 °C)
solubility   0.130 g/l
density   1.77 g/cm3 (20 °C)
flash point   48 °C
autoignition temp.   635 °C
mp   <-20 °C
LD 50 oral   LD50 Rat > 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.775 - 1.785
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H315: Causes skin irritation.
H318: Causes serious eye damage.
H351: Suspected of causing cancer.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P210: Keep away from heat.
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, II
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, II
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, II
Categories of danger 
flammable, irritant, carcinogenic, dangerous for the environment
R Phrase 
R 10-38-40-41-51/53
Flammable.Irritating to skin.Limited evidence of a carcinogenic effect.Risk of serious damage to eyes.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 23-26-36/37/39-61
Do not breathe vapour.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?