• USA Home
  • 8.14967 - 4-Trifluoromethoxyphenol

EMAIL THIS PAGE TO A FRIEND
8.14967 EMD Millipore

4-Trifluoromethoxyphenol

4-Trifluoromethoxyphenol for synthesis. CAS 828-27-3, molar mass 178.11 g/mol., for synthesis

  • CAS Number 828-27-3

  • Linear Formula C₇H₅F₃O₂

  • Molecular Weight 178.11 g/mol

  •  EC Number 212-583-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8149670010)
bp   180 °C (1013 hPa)
density   1.39 g/cm3 (16 °C)
flash point   86 °C
LD 50 oral   LD50 Rat 207 mg/kg
LD 50 dermal   LD50 Rat 2271 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.380 - 1.384
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 50 00
Hazard Statement(s) 
H301: Toxic if swallowed.
H314: Causes severe skin burns and eye damage.
H412: Harmful to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1 C Combustible, acute toxicity cat. 3 / toxic or substances with chronic effects
WGK 
WGK 2 water endangering
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 2922 , 8 (6.1), III
Declaration (transport by air) IATA-DGR 
UN 2922 , 8 (6.1), III
Declaration (transport by sea) IMDG-Code 
UN 2922 , 8 (6.1), III
Categories of danger 
harmful, corrosive, dangerous for the environment
R Phrase 
R 22-34-52/53
Harmful if swallowed.Causes burns.Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 26-36/37/39-45-61
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?