• USA Home
  • 8.14968 - 3,4,5-Trifluorobenzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.14968 EMD Millipore

3,4,5-Trifluorobenzaldehyde

3,4,5-Trifluorobenzaldehyde for synthesis. CAS 132123-54-7, molar mass 160.09 g/mol., for synthesis

  • CAS Number 132123-54-7

  • Linear Formula C₇H₃F₃O

  • Molecular Weight 160.09 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8149680025)
bp   176 - 183 °C (1013 hPa)
vapor pressure   2.8 hPa (20 °C)
solubility   2.2 g/l
density   1.42 g/cm3 (20 °C)
flash point   65 °C
mp   25.3 °C
LD 50 oral   LD50 Rat 200 - 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2913 00 00
Hazard Statement(s) 
H301: Toxic if swallowed.
H317: May cause an allergic skin reaction.
H400: Very toxic to aquatic life.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P273: Avoid release to the environment.
P280: Wear protective gloves.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, II
Categories of danger 
harmful, sensitizing, dangerous for the environment
R Phrase 
R 22-43-50
Harmful if swallowed.May cause sensitisation by skin contact.Very toxic to aquatic organisms.
S Phrase 
S 24-37-61
Avoid contact with skin.Wear suitable gloves.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?