• USA Home
  • 8.14969 - 3,4,5-Trifluorophenol

EMAIL THIS PAGE TO A FRIEND
8.14969 EMD Millipore

3,4,5-Trifluorophenol

3,4,5-Trifluorophenol for synthesis. CAS 99627-05-1, molar mass 148.09 g/mol., for synthesis

  • CAS Number 99627-05-1

  • Linear Formula C₆H₃F₃O

  • Molecular Weight 148.09 g/mol

  •  EC Number 436-120-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8149690001)
solubility   104 g/l OECD Test Guideline 105
density   1.629 g/cm3 (20 °C)
mp   48 °C
LD 50 oral   LD50 Rat 200 - 2000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 45 °C
  Melting range (upper value): ≤ 49 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2908 19 00
Hazard Statement(s) 
H301: Toxic if swallowed.
H312 + H332: Harmful in contact with skin or if inhaled
H314: Causes severe skin burns and eye damage.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Corrosive  Corrosive
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Declaration (railroad and road) ADR, RID 
UN 2923 , 8 (6.1), II
Declaration (transport by air) IATA-DGR 
UN 2923 , 8 (6.1), II
Declaration (transport by sea) IMDG-Code 
UN 2923 , 8 (6.1), II
Categories of danger 
harmful, corrosive, dangerous for the environment
R Phrase 
R 20/21/22-34-51/53
Harmful by inhalation, in contact with skin and if swallowed.Causes burns.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 26-36/37/39-45-61
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?