• USA Home
  • 8.14972 - 2,5-Dimethylanisole

EMAIL THIS PAGE TO A FRIEND
8.14972 EMD Millipore

2,5-Dimethylanisole

2,5-Dimethylanisole for synthesis. CAS 1706-11-2, molar mass 136.2 g/mol., for synthesis

Synonym: 1,4-Dimethyl-2-methoxybenzene

  • CAS Number 1706-11-2

  • Linear Formula C₉H₁₂O

  • Molecular Weight 136.2 g/mol

  •  EC Number 216-943-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8149720025)
density   0.963 g/cm3 (20 °C)
flash point   66 °C
mp   90 - 92 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 30 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?