EMAIL THIS PAGE TO A FRIEND
8.14977 EMD Millipore

Butyramide

Butyramide for synthesis. CAS 541-35-5, molar mass 87.12 g/mol., for synthesis

Synonym: n-Butyramide

  • CAS Number 541-35-5

  • Linear Formula C₄H₉NO

  • Molecular Weight 87.12 g/mol

  •  EC Number 208-776-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8149770100)
solubility   163 g/l
density   1.03 g/cm3
mp   114 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 114 °C
  Melting range (upper value): ≤ 116 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 19 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?