• USA Home
  • 8.14985 - Ethylene glycol tert-butylethyl ether

EMAIL THIS PAGE TO A FRIEND
8.14985 EMD Millipore

Ethylene glycol tert-butylethyl ether

Ethylene glycol tert-butylethyl ether for synthesis. CAS 51422-54-9, pH 7 (H₂O)., for synthesis

  • CAS Number 51422-54-9

  • Linear Formula C₈H₁₈O₂

  • Molecular Weight 146.23 g/mol

  •  EC Number 257-196-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8149850100)
bp   155 °C (1013 hPa)
expl. lim.     0.8 - 8.5 %(V)
vapor pressure   4 hPa
solubility   30 g/l
density   0.83 g/cm3 (20 °C)
flash point   27 °C
autoignition temp.   230 °C
mp   -70 °C
pH   7 (H₂O)
LD 50 oral   LD50 Rat 3600 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Peroxide (as H₂O₂): ≤ 0.005 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 19 90
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P210: Keep away from heat.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
flammable, irritant
R Phrase 
R 10-36/38
Flammable.Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?