• USA Home
  • 8.14991 - Ethyl 2,3-epoxy-3-phenylbutyrate

EMAIL THIS PAGE TO A FRIEND
8.14991 EMD Millipore

Ethyl 2,3-epoxy-3-phenylbutyrate

Ethyl 2,3-epoxy-3-phenylbutyrate for synthesis. CAS 77-83-8, molar mass 206.24 g/mol., for synthesis

Synonym: Ethyl 3-methyl-3-phenylglycidate

  • CAS Number 77-83-8

  • Linear Formula C₁₂H₁₄O₃

  • Molecular Weight 206.24 g/mol

  •  EC Number 201-061-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8149910250)
density   1.092 g/cm3 (20 °C)
LD 50 oral   LD50 Rat 5470 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.092 - 1.096
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 99 90
RTECS 
MW5250000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?