EMAIL THIS PAGE TO A FRIEND
8.16022 EMD Millipore

1-Butylpyrrolidine

1-Butylpyrrolidine for synthesis. CAS 767-10-2, molar mass 127.23 g/mol., for synthesis

  • CAS Number 767-10-2

  • Linear Formula C₈H₁₇N

  • Molecular Weight 127.23 g/mol

  •  EC Number 212-179-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8160220100)
bp   156 °C (1005 hPa)
density   0.818 g/cm3 (20 °C)
flash point   36 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.5 % (a/a)
  Density (d 20 °C/ 4 °C): 0.816 - 0.820
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
RTECS 
UX9800000
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 2929 , 6.1 (3), II
Declaration (transport by air) IATA-DGR 
UN 2929 , 6.1 (3), II
Declaration (transport by sea) IMDG-Code 
UN 2929 , 6.1 (3), II
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?