• USA Home
  • 8.16101 - 4,5-Dicyanoimidazole

EMAIL THIS PAGE TO A FRIEND
8.16101 EMD Millipore

4,5-Dicyanoimidazole

4,5-Dicyanoimidazole for synthesis. CAS 1122-28-7, molar mass 118.1 g/mol., for synthesis

  • CAS Number 1122-28-7

  • Linear Formula C₅H₂N₄

  • Molecular Weight 118.1 g/mol

  •  EC Number 214-344-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8161010025)
mp   171 - 175 °C
LD 50 oral   LD50 Rat 600 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 173 °C
  Melting range (upper value): ≤ 176 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 29 90
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
S Phrase 
S 22-24/25
Do not breathe dust.Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?