• USA Home
  • 8.16117 - 2,4-Dimethylacetophenone

EMAIL THIS PAGE TO A FRIEND
8.16117 EMD Millipore

2,4-Dimethylacetophenone

2,4-Dimethylacetophenone for synthesis. CAS 89-74-7, molar mass 148.21 g/mol., for synthesis

  • CAS Number 89-74-7

  • Linear Formula C₁₀H₁₂O

  • Molecular Weight 148.21 g/mol

  •  EC Number 201-935-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8161170050)
density   0.998 g/cm3 (20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.996 - 1.000
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 39 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?