• USA Home
  • 8.16118 - Ethyl 2-bromobenzoate

EMAIL THIS PAGE TO A FRIEND
8.16118 EMD Millipore

Ethyl 2-bromobenzoate

Ethyl 2-bromobenzoate for synthesis. CAS 6091-64-1, molar mass 229.07 g/mol., for synthesis

Synonym: 2-Bromobenzoic acid ethyl ester

  • CAS Number 6091-64-1

  • Linear Formula C₉H₉BrO₂

  • Molecular Weight 229.07 g/mol

  •  EC Number 228-034-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8161180025)
density   1.441 g/cm3 (20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.441 - 1.445
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?