• USA Home
  • 8.16120 - Methyl 2-chlorobenzoate

EMAIL THIS PAGE TO A FRIEND
8.16120 EMD Millipore

Methyl 2-chlorobenzoate

Methyl 2-chlorobenzoate for synthesis. CAS 610-96-8, molar mass 170.6 g/mol., for synthesis

Synonym: 2-Chlorobenzoic acid methyl ester

  • CAS Number 610-96-8

  • Linear Formula C₈H₇ClO₂

  • Molecular Weight 170.6 g/mol

  •  EC Number 210-242-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8161200025)
density   1.19 g/cm3 (25 °C)
flash point   109 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.244 - 1.250
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?