• USA Home
  • 8.16122 - O-(1H-6-Chlorobenzotriazol-1-yl)-1,1,3,3- tetramethyluronium hexafluorophosphate

EMAIL THIS PAGE TO A FRIEND
8.16122 EMD Millipore

O-(1H-6-Chlorobenzotriazol-1-yl)-1,1,3,3- tetramethyluronium hexafluorophosphate

O-(1H-6-Chlorobenzotriazol-1-yl)-1,1,3,3- tetramethyluronium hexafluorophosphate for synthesis. CAS 330645-87-9, molar mass 413.69 g/mol., for synthesis

Synonym: HCTU

  • CAS Number 330645-87-9

  • Linear Formula C₁₁H₁₅ClF₆N₅OP

  • Molecular Weight 413.69 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8161220025)
mp   >185 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Water (K. F.): ≤ 0.50 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Hazard Statement(s) 
H228: Flammable solid.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
4.1B Flammable solid hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Symbols 
Flammable  Flammable
Declaration (railroad and road) ADR, RID 
UN 1325 , 4.1, II
Declaration (transport by air) IATA-DGR 
UN 1325 , 4.1, II
Declaration (transport by sea) IMDG-Code 
UN 1325 , 4.1, II
Categories of danger 
highly flammable
R Phrase 
R 11
Highly flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?