• USA Home
  • 8.18000 - Toluene 4-sulfonic acid silver salt

EMAIL THIS PAGE TO A FRIEND
8.18000 EMD Millipore

Toluene 4-sulfonic acid silver salt

Toluene 4-sulfonic acid silver salt for synthesis. CAS No. 16836-95-6, EC Number 240-859-0., for synthesis

Synonym: Silver tosylate, Silver 4-toluenesulfonate

  • CAS Number 16836-95-6

  • Linear Formula C₇H₇O₃S*Ag

  • Molecular Weight 279.07 g/mol

  •  EC Number 240-859-0

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds
packaging   10 g in Glass bottle (8180000010)
storage temp.   Store below +30°C.
specifications   Assay (from Ag): ≥ 99.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2843 29 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 1 slightly water endangering
Disposal 
27
Residues containing valuable recoverable metals should be forwarded for recycling. Container H.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 1759 , 8, III
Declaration (transport by air) IATA-DGR 
UN 1759 , 8, III
Declaration (transport by sea) IMDG-Code 
UN 1759 , 8, III
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
S Phrase 
S 26-37/39
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable gloves and eye/face protection.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?