• USA Home
  • 8.18027 - Benzophenone hydrazone

EMAIL THIS PAGE TO A FRIEND
8.18027 EMD Millipore

Benzophenone hydrazone

Benzophenone hydrazone for synthesis. CAS No. 5350-57-2, EC Number 226-321-8., for synthesis

  • CAS Number 5350-57-2

  • Linear Formula C₁₃H₁₂N₂

  • Molecular Weight 196.25 g/mol

  •  EC Number 226-321-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Glass bottle (8180270050)
bp   225 - 230 °C (73 hPa)
flash point   146 °C
mp   96 - 99 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Melting range (lower value): ≥ 96 °C
  Melting range (upper value): ≤ 99 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2928 00 90
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?