• USA Home
  • 8.18032 - Ethyl 1,3-dithiane-2-carboxylate

EMAIL THIS PAGE TO A FRIEND
8.18032 EMD Millipore

Ethyl 1,3-dithiane-2-carboxylate

Ethyl 1,3-dithiane-2-carboxylate for synthesis. CAS 20462-00-4, molar mass 192.29 g/mol., for synthesis

Synonym: 1,3-Dithiane-2-carboxylic acid ethyl ester

  • CAS Number 20462-00-4

  • Linear Formula C₇H₁₂O₂S₂

  • Molecular Weight 192.29 g/mol

  •  EC Number 243-838-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8180320005)
density   1.22 g/cm3 (20 °C)
mp   19 - 21 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Purity (GC): ≥ 97
  Identity (IR-spectrum): conforms
  Density (20/4 °C): 1.218 - 1.220

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
6
Sulfides and sulfites as well as other reducing substances, inorganic cyanides and isonitriles: Stir into sodium hypochlorite solution (Cat. No. 105614) and, if necessary, leave to react for several days. Caution: some substances may show a violent reaction! Draw off any toxic or flammable gases that may be formed. Excess oxidant should be neutralized with sodium thiosulfate (Cat. No. 106513). Container D or E.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?