• USA Home
  • 8.18123 - Cyclopropylmethanol

EMAIL THIS PAGE TO A FRIEND
8.18123 EMD Millipore

Cyclopropylmethanol

Cyclopropylmethanol for synthesis. CAS 2516-33-8, EC Number 219-735-5, molar mass 72.1 g/mol., for synthesis

Synonym: Cyclopropyl carbinol, Hydroxymethylcyclopropane

  • CAS Number 2516-33-8

  • Linear Formula C₄H₈O

  • Molecular Weight 72.1 g/mol

  •  EC Number 219-735-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8181230025)
bp   122 - 126 °C (1013 hPa)
density   0.904 g/cm3 (20 °C)
flash point   35 °C
autoignition temp.   310 °C
mp   <-60 °C
shipped in   ambient
storage temp.   Storage temperature: no restrictions.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.911 - 0.914
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 19 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
RTECS 
GZ2212250
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1987 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1987 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1987 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?