• USA Home
  • 8.18158 - Bis(triphenylphosphine)nickel(II) chloride

EMAIL THIS PAGE TO A FRIEND
8.18158 EMD Millipore

Bis(triphenylphosphine)nickel(II) chloride

Bis(triphenylphosphine)nickel(II) chloride for synthesis. CAS No. 14264-16-5, EC Number 238-154-8., for synthesis

Synonym: Dichlorobis(triphenylphosphine)nickel(II), Nickel bis(triphenylphosphine)dichloride

  • CAS Number 14264-16-5

  • Linear Formula C₃₆H₃₀Cl₂NiP₂

  • Molecular Weight 654.18 g/mol

  •  EC Number 238-154-8

Purchase

Properties

packaging   10 g in Glass bottle (8181580010)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
15
Heavy metal-containing solutions and solids: container E. Stir Raney nickel (also: Urushibara nickel) in the form of an aqueous suspension into hydrochloric acid (Cat. No. 100312) until dissolved (container E). Neither Raney nickel itself nor any filter residues should be allowed to dry out, otherwise they will spontaneously ignite in air. In this context, heavy metal means any compound of antimony, arsenic, cadmium, chromium(VI), copper, lead, nickel and tin, as well as these subtances in metallic form, if they are classified as hazardous (acc. to AbfallverzeichnisV - Waste Catalogue Ordinance, Appendix 3). Other heavy metals should be collected separately.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?