• USA Home
  • 8.18159 - 3-Nitrobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.18159 EMD Millipore

3-Nitrobenzyl alcohol

3-Nitrobenzyl alcohol for synthesis. CAS No. 619-25-0, EC Number 210-588-2., for synthesis

  • CAS Number 619-25-0

  • Linear Formula C₇H₇NO₃

  • Molecular Weight 153.14 g/mol

  •  EC Number 210-588-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8181590025)
bp   170 °C (8 hPa)
density   1.30 g/cm3 (20 °C)
flash point   >110 °C
mp   30 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?