• USA Home
  • 8.18170 - 4-Methylmorpholine-4-oxide monohydrate

EMAIL THIS PAGE TO A FRIEND
8.18170 EMD Millipore

4-Methylmorpholine-4-oxide monohydrate

4-Methylmorpholine-4-oxide monohydrate for synthesis. CAS 70187-32-5, molar mass 135.16 g/mol., for synthesis

Synonym: NMO

  • CAS Number 70187-32-5

  • Linear Formula C₅H₁₁NO₂ * H₂O

  • Molecular Weight 135.16 g/mol

  •  EC Number 231-391-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8181700025)
mp   70 - 76 °C
storage temp.   Store below +30°C.
specifications   Assay (HClO₄, calc. on anhydrous substance): ≥ 97.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 99 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?