• USA Home
  • 8.18172 - 2-Chloro-1-methylpyridinium iodide

EMAIL THIS PAGE TO A FRIEND
8.18172 EMD Millipore

2-Chloro-1-methylpyridinium iodide

2-Chloro-1-methylpyridinium iodide for synthesis. CAS 14338-32-0, chemical formula 2-(Cl)-1-(CH₃)C₅H₃NiHI., for synthesis

  • CAS Number 14338-32-0

  • Linear Formula C₆H₇ClIN

  • Molecular Weight 255.49 g/mol

  •  EC Number 238-288-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8181720025)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 99.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 39 99
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?