• USA Home
  • 8.18176 - Diethyl cyanomethylphosphonate

EMAIL THIS PAGE TO A FRIEND
8.18176 EMD Millipore

Diethyl cyanomethylphosphonate

Diethyl cyanomethylphosphonate for synthesis. CAS 2537-48-6, chemical formula NCCH₂P(=O) (OC₂H₅)₂., for synthesis

Synonym: Cyanomethanephosphonic acid diethyl ester

  • CAS Number 2537-48-6

  • Linear Formula C₆H₁₂NO₃P

  • Molecular Weight 177.14 g/mol

  •  EC Number 219-806-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8181760010)
density   1.1 g/cm3 (25 °C)
flash point   113 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.126 - 1.128
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?