• USA Home
  • 8.18177 - Formylmethyltriphenylphosphonium chloride

EMAIL THIS PAGE TO A FRIEND
8.18177 EMD Millipore

Formylmethyltriphenylphosphonium chloride

Formylmethyltriphenylphosphonium chloride for synthesis. CAS 62942-43-2, molar mass 340.78 g/mol., for synthesis

Synonym: Triphenylformylmethylphosphonium chloride

  • CAS Number 62942-43-2

  • Linear Formula C₂₀H₁₈ClOP

  • Molecular Weight 340.78 g/mol

  •  EC Number 263-767-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8181770025)
mp   213 - 217 °C (External MSDS)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 98.0 %
  Melting range (lower value): ≥ 205 °C
  Melting range (upper value): ≤ 210 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 39 90
Hazard Statement(s) 
H302: Harmful if swallowed.
H318: Causes serious eye damage.
Precautionary Statement(s) 
P280: Wear eye protection.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P313: Get medical advice/ attention.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
R Phrase 
R 22-41
Harmful if swallowed.Risk of serious damage to eyes.
S Phrase 
S 26-39
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear eye/face protection.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?