• USA Home
  • 8.18181 - Bis(benzonitrile)-palladium(II) chloride (27.7% Pd)

EMAIL THIS PAGE TO A FRIEND
8.18181 EMD Millipore

Bis(benzonitrile)-palladium(II) chloride (27.7% Pd)

Bis(benzonitrile)-palladium(II) chloride (27.7% Pd) for synthesis. CAS No. 14220-64-5, EC Number 238-085-3., for synthesis

Synonym: Dichlorobis(benzonitrile)palladium(II)

  • CAS Number 14220-64-5

  • Linear Formula C₁₄H₁₀Cl₂N₂Pd

  • Molecular Weight 383.57 g/mol

  •  EC Number 238-085-3

Purchase

Properties

packaging   1 g in Glass bottle (8181810001)
mp   131 °C (External MSDS)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 98.0 %
  Melting range (lower value): ≥ 125 °C
  Melting range (upper value): ≤ 128 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2843 90 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
27
Residues containing valuable recoverable metals should be forwarded for recycling. Container H.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?