• USA Home
  • 8.18207 - (1S)-(-)-Camphanic acid

EMAIL THIS PAGE TO A FRIEND
8.18207 EMD Millipore

(1S)-(-)-Camphanic acid

(1S)-(-)-Camphanic acid for synthesis. CAS 13429-83-9, molar mass 198.21 g/mol., for synthesis

  • CAS Number 13429-83-9

  • Linear Formula C₁₀H₁₄O₄

  • Molecular Weight 198.21 g/mol

  •  EC Number 236-552-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8182070005)
mp   202 - 205 °C
bulk density   430 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 99.0 %
  Melting range (lower value): ≥ 202 °C
  Melting range (upper value): ≤ 205 °C
  Water (K. F.): ≤ 1.0 %
  Spec. rotation [α²0/D (c=1 in dioxan): -21.0 - -19.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 20 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?