• USA Home
  • 8.18223 - 9-Bromophenanthrene

EMAIL THIS PAGE TO A FRIEND
8.18223 EMD Millipore

9-Bromophenanthrene

9-Bromophenanthrene for synthesis. CAS 573-17-1, molar mass 257.12 g/mol., for synthesis

  • CAS Number 573-17-1

  • Linear Formula C₁₄H₉Br

  • Molecular Weight 257.12 g/mol

  •  EC Number 209-351-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   190 °C (2 hPa)
mp   59 - 64 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 62 °C
  Melting range (upper value): ≤ 66 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?