• USA Home
  • 8.18225 - (2R,3R)-(-)-Dimethyl-2,3-O-isopropylidene tartrate

EMAIL THIS PAGE TO A FRIEND
8.18225 EMD Millipore

(2R,3R)-(-)-Dimethyl-2,3-O-isopropylidene tartrate

(2R,3R)-(-)-Dimethyl-2,3-O-isopropylidene tartrate for synthesis. CAS 37031-29-1, molar mass 218.2 g/mol., for synthesis

Synonym: 4R-trans-Dimethyl 2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate, 2,3-O-Isopropylidene-L-tartaric acid dimethyl ester

  • CAS Number 37031-29-1

  • Linear Formula C₉H₁₄O₆

  • Molecular Weight 218.2 g/mol

  •  EC Number 609-324-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
density   1.19 g/cm3 (20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.193 - 1.195
  Spec. rotation [α²0/D (undiluted): -64.0 - -62.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?