• USA Home
  • 8.18226 - Tetrahydro-4H-pyran-4-one

EMAIL THIS PAGE TO A FRIEND
8.18226 EMD Millipore

Tetrahydro-4H-pyran-4-one

Tetrahydro-4H-pyran-4-one for synthesis. CAS 29943-42-8, molar mass 100.11 g/mol., for synthesis

Synonym: Tetrahydro-gamma-pyrone

  • CAS Number 29943-42-8

  • Linear Formula C₅H₈O₂

  • Molecular Weight 100.11 g/mol

  •  EC Number 249-967-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 ml in Glass bottle (8182260001)
density   1.08 g/cm3 (20 °C)
flash point   56 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.083 - 1.085
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?