• USA Home
  • 8.18228 - Benzylammonium chloride

EMAIL THIS PAGE TO A FRIEND
8.18228 EMD Millipore

Benzylammonium chloride

Benzylammonium chloride for synthesis. CAS No. 3287-99-8, EC Number 221-943-6., for synthesis

Synonym: Benzylamine hydrochloride

  • CAS Number 3287-99-8

  • Linear Formula C₇H₁₀ClN

  • Molecular Weight 143.61 g/mol

  •  EC Number 221-943-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8182280100)
solubility   506 g/l (experimental)
autoignition temp.   260 °C
mp   262 - 263 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 98.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 49 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
No Dangerous Good
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?