• USA Home
  • 8.18231 - (S)-(+)-1-Amino-2-propanol

EMAIL THIS PAGE TO A FRIEND
8.18231 EMD Millipore

(S)-(+)-1-Amino-2-propanol

(S)-(+)-1-Amino-2-propanol for synthesis. CAS 2799-17-9, chemical formula H₂NCH₂CH(OH)CH₃., for synthesis

Synonym: S-(+)-Isopropanolamine

  • CAS Number 2799-17-9

  • Linear Formula C₃H₉NO

  • Molecular Weight 75.11 g/mol

  •  EC Number 220-533-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   1 g in Glass bottle (8182310001)
bp   158 - 162 °C (1013 hPa)
vapor pressure   6.7 hPa (20 °C)
density   0.96 g/cm3 (20 °C)
flash point   71 °C
autoignition temp.   335 °C
mp   20 - 23 °C
LD 50 oral   LD50 Rat 4260 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 20 °C
  Melting range (upper value): ≤ 23 °C
  Spec. rotation [α²0/D (c=2 in water): +17.0 - +18.0 °
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 19 00
Hazard Statement(s) 
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 2735 , 8, II
Declaration (transport by air) IATA-DGR 
UN 2735 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 2735 , 8, II, Segregation Group: 18 (Alkalis)
Categories of danger 
corrosive
R Phrase 
R 34
Causes burns.
S Phrase 
S 23-26-36-45
Do not breathe vapour.In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?