• USA Home
  • 8.18234 - (+)-5,6-O-Isopropylidene-L-ascorbic acid

EMAIL THIS PAGE TO A FRIEND
8.18234 EMD Millipore

(+)-5,6-O-Isopropylidene-L-ascorbic acid

(+)-5,6-O-Isopropylidene-L-ascorbic acid for synthesis. CAS 15042-01-0, molar mass 216.19 g/mol., for synthesis

  • CAS Number 15042-01-0

  • Linear Formula C₉H₁₂O₆

  • Molecular Weight 216.19 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8182340010)
mp   223 - 226 °C (decomposition)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (iodometric): ≥ 98.0 %
  Spec. rotation [α²0/D (c=5 in methanol): +10.0 - +13.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 20 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?