• USA Home
  • 8.18239 - (-)-1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose

EMAIL THIS PAGE TO A FRIEND
8.18239 EMD Millipore

(-)-1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose

(-)-1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose for synthesis. CAS 582-52-5, molar mass 260.29 g/mol., for synthesis

Synonym: Diacetoneglucose

  • CAS Number 582-52-5

  • Linear Formula C₁₂H₂₀O₆

  • Molecular Weight 260.29 g/mol

  •  EC Number 209-486-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
mp   106 - 109 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 106 °C
  Melting range (upper value): ≤ 109 °C
  Spec. rotation [α²0/D (c=1 in chloroform): -12.0 - -11.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2940 00 00
RTECS 
LZ4958000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?