• USA Home
  • 8.18258 - Diethyl chloromalonate

EMAIL THIS PAGE TO A FRIEND
8.18258 EMD Millipore

Diethyl chloromalonate

Diethyl chloromalonate for synthesis. CAS 14064-10-9, chemical formula ClCH(COOC₂H₅)₂., for synthesis

Synonym: Chloromalonic acid diethyl ester

  • CAS Number 14064-10-9

  • Linear Formula C₇H₁₁ClO₄

  • Molecular Weight 194.61 g/mol

  •  EC Number 237-913-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8182580100)
bp   221 - 222 °C
density   1.18 g/cm3 (20 °C)
flash point   113 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 89.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 19 80
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
H400: Very toxic to aquatic life.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 3082 , 9, III
Declaration (transport by air) IATA-DGR 
UN 3082 , 9, III
Declaration (transport by sea) IMDG-Code 
UN 3082 , 9, III, Marine Pollutant: P
Categories of danger 
irritant, dangerous for the environment
R Phrase 
R 36/37/38-50
Irritating to eyes, respiratory system and skin.Very toxic to aquatic organisms.
S Phrase 
S 61
Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?