• USA Home
  • 8.18312 - 3-Nitro-1H-1,2,4-triazole

EMAIL THIS PAGE TO A FRIEND
8.18312 EMD Millipore

3-Nitro-1H-1,2,4-triazole

3-Nitro-1H-1,2,4-triazole for synthesis. CAS 24807-55-4, molar mass 114.06 g/mol., for synthesis

  • CAS Number 24807-55-4

  • Linear Formula C₂H₂N₄O₂

  • Molecular Weight 114.06 g/mol

  •  EC Number 246-468-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8183120005)
mp   210 - 215 °C
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 210 °C
  Melting range (upper value): ≤ 215 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?