• USA Home
  • 8.18317 - (2S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine

EMAIL THIS PAGE TO A FRIEND
8.18317 EMD Millipore

(2S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine

(2S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine for synthesis. CAS 78342-42-4, molar mass 184.23 g/mol., for synthesis

Synonym: Schoellkopf-Hartwig-Reagent, Bislactimether

  • CAS Number 78342-42-4

  • Linear Formula C₉H₁₆N₂O₂

  • Molecular Weight 184.23 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   103 - 104 °C (20 hPa)
density   1.03 g/cm3 (20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.027 - 1.029
  Spec. rotation [α²0/D (c=1 in ethanol): +105.0 - +109.0 °
  Identity (IR): passes test
  enantiomeric excess (NMR-spectrum): ≥ 97.0 %

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?