• USA Home
  • 8.18351 - 2,3-Dimethoxybenzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.18351 EMD Millipore

2,3-Dimethoxybenzaldehyde

2,3-Dimethoxybenzaldehyde for synthesis. CAS 86-51-1, chemical formula 2,3-(CH₃O)₂C₆H₃CHO., for synthesis

  • CAS Number 86-51-1

  • Linear Formula C₉H₁₀O₃

  • Molecular Weight 166.17 g/mol

  •  EC Number 201-677-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8183510025)
bp   132 - 137 °C (16 hPa)
flash point   113 °C
mp   51 - 54 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 49 °C
  Melting range (upper value): ≤ 53 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2912 49 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?