• USA Home
  • 8.18353 - 3,5-Dimethoxy-4-hydroxybenzaldehyde

EMAIL THIS PAGE TO A FRIEND
8.18353 EMD Millipore

3,5-Dimethoxy-4-hydroxybenzaldehyde

3,5-Dimethoxy-4-hydroxybenzaldehyde for synthesis. CAS No. 134-96-3, EC Number 205-167-5., for synthesis

Synonym: Syringaldehyde

  • CAS Number 134-96-3

  • Linear Formula C₉H₁₀O₄

  • Molecular Weight 182.17 g/mol

  •  EC Number 205-167-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8183530010)
bp   192 - 193 °C (19 hPa)
mp   109 - 112 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 109 °C
  Melting range (upper value): ≤ 112 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2912 49 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?