• USA Home
  • 8.18365 - 2-Aminobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.18365 EMD Millipore

2-Aminobenzyl alcohol

2-Aminobenzyl alcohol for synthesis. CAS 5344-90-1, chemical formula 2-(NH₂)C₆H₄CH₂OH., for synthesis

Synonym: 2-(Hydroxymethyl)aniline

  • CAS Number 5344-90-1

  • Linear Formula C₇H₉NO

  • Molecular Weight 123.15 g/mol

  •  EC Number 226-293-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8183650010)
bp   159 - 161 °C (13 hPa)
mp   81 - 83 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 81 °C
  Melting range (upper value): ≤ 84 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 19 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?