EMAIL THIS PAGE TO A FRIEND
8.18366 EMD Millipore

N-Acetylglycine

N-Acetylglycine for synthesis. CAS 543-24-8, EC Number 208-839-6, chemical formula CH₃CONHCH₂COOH., for synthesis

Synonym: Aceturic acid

  • CAS Number 543-24-8

  • Linear Formula C₄H₇NO₃

  • Molecular Weight 117.1 g/mol

  •  EC Number 208-839-6

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8183660100)
bp   >300 °C (1013 hPa)
solubility   26.3 g/l
mp   206 - 209 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 99.0 %
  Melting range (lower value): ≥ 206 °C
  Melting range (upper value): ≤ 209 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 19 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?