• USA Home
  • 8.18371 - α,α,α-Trifluoroacetophenone

EMAIL THIS PAGE TO A FRIEND
8.18371 EMD Millipore

α,α,α-Trifluoroacetophenone

a,a,a-Trifluoroacetophenone for synthesis. CAS 434-45-7, EC Number 207-103-1, chemical formula C₆H₅-C(O)-CF₃., for synthesis

Synonym: Trifluoroacetylbenzene

  • CAS Number 434-45-7

  • Linear Formula C₈H₅F₃O

  • Molecular Weight 174.12 g/mol

  •  EC Number 207-103-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   165 °C (1013 hPa)
density   1.28 g/cm3 (20 °C)
flash point   41 °C
mp   -40 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.278 - 1.279
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 79 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 1224 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1224 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1224 , 3, III
Categories of danger 
flammable, irritant
R Phrase 
R 10-36/37/38
Flammable.Irritating to eyes, respiratory system and skin.
S Phrase 
S 26
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?