• USA Home
  • 8.18373 - Tris[2-(2-methoxyethoxy)ethyl]amine

EMAIL THIS PAGE TO A FRIEND
8.18373 EMD Millipore

Tris[2-(2-methoxyethoxy)ethyl]amine

Tris[2-(2-methoxyethoxy)ethyl]amine for synthesis. CAS 70384-51-9, EC Number 274-590-5, chemical formula N(CH₂CH₂OCH₂CH₂OCH₃)₃., for synthesis

  • CAS Number 70384-51-9

  • Linear Formula C₁₅H₃₃NO₆

  • Molecular Weight 323.42 g/mol

  •  EC Number 274-590-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8183730025)
bp   329 - 331 °C (1013 hPa)
density   1.01 g/cm3 (20 °C)
flash point   163 °C
autoignition temp.   240 °C
mp   <-100 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 94.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.008 - 1.012
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 19 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
S Phrase 
S 24
Avoid contact with skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?