• USA Home
  • 8.18375 - Bis(3,5,5-trimethylhexyl) phthalate

EMAIL THIS PAGE TO A FRIEND
8.18375 EMD Millipore

Bis(3,5,5-trimethylhexyl) phthalate

Bis(3,5,5-trimethylhexyl) phthalate for synthesis. CAS 14103-61-8, pH 7 (H₂O, 20 °C)., for synthesis

Synonym: Phthalic acid bis(3,5,5-trimethylhexyl) ester

  • CAS Number 14103-61-8

  • Linear Formula C₂₆H₄₂O₄

  • Molecular Weight 418.61 g/mol

  •  EC Number 237-954-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8183750100)
bp   235 - 238 °C
expl. lim.     0.4 - 2.9 %(V)
vapor pressure   <0.001 hPa (20 °C)
density   0.98 g/cm3 (20 °C)
flash point   226 °C
autoignition temp.   305 °C
mp   -49 °C
pH   7 (H₂O, 20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 99.0 %
  Density (d 20 °C/ 4 °C): 0.973 - 0.977
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 34 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?