• USA Home
  • 8.18379 - 4-Aminobenzophenone

EMAIL THIS PAGE TO A FRIEND
8.18379 EMD Millipore

4-Aminobenzophenone

4-Aminobenzophenone for synthesis. CAS 1137-41-3, molar mass 197.23 g/mol., for synthesis

Synonym: 4-Benzoylaniline

  • CAS Number 1137-41-3

  • Linear Formula C₁₃H₁₁NO

  • Molecular Weight 197.23 g/mol

  •  EC Number 214-506-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Plastic bottle (8183790005)
bp   245 - 247 °C (17 hPa)
mp   118 - 121 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.5 % (a/a)
  Melting range (lower value): ≥ 118 °C
  Melting range (upper value): ≤ 121 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 39 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?