• USA Home
  • 8.18396 - Diethyl isopropylidenemalonate

EMAIL THIS PAGE TO A FRIEND
8.18396 EMD Millipore

Diethyl isopropylidenemalonate

Diethyl isopropylidenemalonate for synthesis. CAS 6802-75-1, chemical formula (CH₃)₂C=C(COOC₂H₅)₂., for synthesis

Synonym: Isopropylidenemalonic acid diethyl ester

  • CAS Number 6802-75-1

  • Linear Formula C₁₀H₁₆O₄

  • Molecular Weight 200.23 g/mol

  •  EC Number 229-876-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   64 - 86 °C (24 hPa)
density   1.03 g/cm3 (20 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.024 - 1.026
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 19 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?