• USA Home
  • 8.18397 - Isosorbide dimethyl ether

EMAIL THIS PAGE TO A FRIEND
8.18397 EMD Millipore

Isosorbide dimethyl ether

Isosorbide dimethyl ether for synthesis. CAS 5306-85-4, pH 7.0 - 8.0 (100 g/l, H₂O, 20 °C)., for synthesis

Synonym: 1,4:3,6-Dianhydrosorbitol 2,5-dimethyl ether, 1,4:3,6-Dianhydro-2,5-O-dimethyl-D-glucitol

  • CAS Number 5306-85-4

  • Linear Formula C₈H₁₄O₄

  • Molecular Weight 174.19 g/mol

  •  EC Number 226-159-8

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, Chemical Synthesis, Ethers, Organic Building Blocks,
bp   93 - 95 °C (0.20 hPa)
density   1.17 g/cm3 (20 °C)
flash point   103 °C
mp   -70 °C
pH   7.0 - 8.0 (100 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 6500 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.165 - 1.168
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?