• USA Home
  • 8.18398 - Perfluorobutyl iodide

EMAIL THIS PAGE TO A FRIEND
8.18398 EMD Millipore

Perfluorobutyl iodide

Perfluorobutyl iodide (stabilised with silver platelets) for synthesis. CAS 423-39-2, EC Number 207-025-8, chemical formula CF₃(CF₂)₂CF₂I., (stabilised with silver platelets) for synthesis

Synonym: 1,1,1,2,2,3,3,4,4,-Nonafluoro-4-iodobutane

  • CAS Number 423-39-2

  • Linear Formula C₄F₉I

  • Molecular Weight 345.93 g/mol

  •  EC Number 207-025-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8183980010)
bp   64 - 66 °C (973 hPa)
vapor pressure   133 hPa (19 °C)
density   2.05 g/cm3 (20 °C)
flash point   >115 °C
mp   -89 - -87 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 2.050 - 2.051
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 78 00
Hazard Statement(s) 
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P273: Avoid release to the environment.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Declaration (railroad and road) ADR, RID 
UN 3082 , 9, III
Declaration (transport by air) IATA-DGR 
UN 3082 , 9, III
Declaration (transport by sea) IMDG-Code 
UN 3082 , 9, III, Marine Pollutant: P
Categories of danger 
dangerous for the environment
R Phrase 
R 51/53
Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 24/25-61
Avoid contact with skin and eyes.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?