• USA Home
  • 8.18399 - Perfluorohexyl iodide

EMAIL THIS PAGE TO A FRIEND
8.18399 EMD Millipore

Perfluorohexyl iodide

Perfluorohexyl iodide for synthesis. CAS 355-43-1, EC Number 206-586-6, chemical formula CF₃(CF₂)₄CF₂I., for synthesis

Synonym: 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-6-iodohexane

  • CAS Number 355-43-1

  • Linear Formula C₆F₁₃I

  • Molecular Weight 445.94 g/mol

  •  EC Number 206-586-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
vapor pressure   133 hPa (60 °C)
density   2.06 g/cm3 (25 °C)
flash point   115 °C (External MSDS)
mp   -46 °C (Lit.)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 2.048 - 2.052
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 78 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
S Phrase 
S 24/25
Avoid contact with skin and eyes.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?