• USA Home
  • 8.18403 - N-(Trimethylsilyl)acetamide

EMAIL THIS PAGE TO A FRIEND
8.18403 EMD Millipore

N-(Trimethylsilyl)acetamide

N-(Trimethylsilyl)acetamide for synthesis. CAS 13435-12-6, EC Number 236-565-7, chemical formula CH₃CONHSi(CH₃)₃., for synthesis

Synonym: TMS-acetamide

  • CAS Number 13435-12-6

  • Linear Formula C₅H₁₃NOSi

  • Molecular Weight 131.24 g/mol

  •  EC Number 236-565-7

Purchase

Properties

packaging   25 g in Glass bottle (8184030025)
bp   104 - 106 °C (1013 hPa)
flash point   70 °C
autoignition temp.   400 °C
mp   40 - 45 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
AD0250000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?